missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 47-200 Light Duty Nitrile Coated Gloves

Catalog No. 18999454B
Change view
Click to view available options
:
8.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18999454B 8.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999454B Supplier Ansell Supplier No. 4720011080
Only null left

Gloves are compliant with FDA food handling requirements and feature a lightweight nitrile coating that repels oil and grease.

  • Coolness and comfort of cotton, but with enhanced grip capabilities
  • Lightweight nitrile coating repels oil and grease
  • Coating only on fingers and palm to protect the working area, while still allowing the hand to breathe
  • Ergonomically designed to be cool, comfortable, and flexible
  • Compliant with FDA food handling requirements

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Coated Area Palm, Fingers
Construction Cut and Sewn
Coating Material Nitrile
Color Green
Cut Resistance Rating ANSI A1
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 8.5
For Use With (Application) Automotive Assembly, Machinery/Equipment, Metal Fabrication, and Chemical Handling
Glove Cuff Color Natural
Glove Cuff Style Knit
Series 47-200
Show More Show Less