missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Chemical- and Flame-Resistant Bib Pants

Catalog No. 17987449d
Change view
Click to view available options
Color:
Red
Material:
CPC Polyester
Size:
Large
Size (English) Waist:
52 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Size Size (English) Waist
17987449D Red CPC Polyester Large 52 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.
Catalog No. 17987449D Supplier Ansell™ Supplier No. 66662L
Only null left
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.

Lightweight and breathable CPC garments made with Gore™ ChemSplash™ fabric reduce overheating and fatigue while preventing liquid chemical splash penetration

Gore membrane is a bicomponent PTFE membrane that prevents liquid penetration but allows a vapor permeation, allowing perspiration to evaporate and cool the wearer.

  • Garments stitched with Gore thread
  • All seams are heat and pressure sealed with Gore-Seam™ tape to prevent liquid penetration
  • Fabrics are durable, flexible, tear and snag resistant, washable, and reusable

Coveralls
  • Feature two-way, heavy-duty, static-free zipper
  • Double storm flap; stand-up collar; raglan sleeves
  • Hook-and-loop closures on storm flap, collar, sleeves, and leg bottoms
Jackets
  • Heavy-duty zipper
  • Double storm flap; stand-up collar; raglan sleeves
  • Hook-and-loop closures on storm flap, collar, and sleeves
  • Sized to be worn over clothing
Bib Pants
  • Elastic suspenders with quick-release clips for fast removal
  • Leg bottoms have hook-and-loop closures
Regular Hoods
  • Attach, by means of a hook-and-loop fastener, to back of collar on coveralls or jacket
  • Drawstring closure gives snug fit over hardhat

Order Info

For chemical splash protection, choose CPC garments made with red polyester Gore ChemSplash fabric (7 oz. per sq. yd.). When flame protection is also required, choose garments made with blue aramid (Nomex™) Gore ChemSplash fabric (6 oz. per sq. yd.); when exposed to flame, it will self-extinguish when removed from the flame source, and not contribute to burn injury.

Specifications

Closure Type Double Front Flap (Hook and Loop)
Color Red
Material CPC Polyester
Size Large
Suspender Elastic
Construction Trilaminate
For Use With (Application) Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals
Length (English) Inseam 31 in.
Length (Metric) Inseam 79 cm
Size (English) Waist 52 in.
Certifications/Compliance NFPA, ASTM F903
Show More Show Less