missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin V Recombinant Protein, FITC, eBioscience™
Recombinant Protein
Supplier: Invitrogen™ BMS306FI100
Description
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.Chemical Identifiers
| P08758, P14668, P48036 | |
| Neutralization, Flow Cytometry | |
| 11747, 25673, 308 | |
| Annexin V | |
| Purified | |
| 4°C, store in dark, DO NOT FREEZE! | |
| E. Coli | |
| Anxa5 | |
| Protein | |
| FITC | |
| E. coli | |
| Liquid | |
| >89% by SDS-PAGE and RP-HPLC |
| 5 μL/Test | |
| TBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| 35.8kDa | |
| 7.4 | |
| E. coli | |
| RUO | |
| Annexin V | |
| Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS). | |
| All | |
| All | |
| Recombinant | |
| Recombinant |
Specifications
| P08758, P14668, P48036 | |
| 5 μL/Test | |
| TBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| 35.8kDa | |
| 7.4 | |
| E. coli | |
| RUO | |
| Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha | |
| Anxa5 | |
| Protein | |
| FITC | |
| E. coli | |
| E. coli | |
| Recombinant |
| 100 Tests | |
| Neutralization, Flow Cytometry | |
| 11747, 25673, 308 | |
| Annexin V | |
| Purified | |
| 4°C, store in dark, DO NOT FREEZE! | |
| E. Coli | |
| Annexin V | |
| Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS). | |
| All | |
| All | |
| Recombinant | |
| Liquid | |
| >89% by SDS-PAGE and RP-HPLC |
Safety and Handling
ShelfLife : 12 Months