missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Annexin V Conjugates for Apoptosis Detection

Catalog No. A13201
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Excitation/Emission:
410/455
494/518
495/519
555/565
565/578
578/603
590/617
650/665
679/702
Flow Cytometer Laser Lines:
405
488
488, 532, 561
532
532, 561
633-637
Conjugate:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 568
Alexa Fluor 594
Alexa Fluor 647
Alexa Fluor 680
Biotin-X
FITC
Pacific Blue
PE
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Excitation/Emission Flow Cytometer Laser Lines Conjugate
A13201 495/519 488 Alexa Fluor 488
A13199 494/518 488 FITC
501121497 578/603 532, 561 Alexa Fluor 568
A13203 590/617 532 Alexa Fluor 594
501121498 - - Biotin-X
A23204 650/665 633-637 Alexa Fluor 647
A35108 555/565 532, 561 Alexa Fluor 555
501121504 679/702 633-637 Alexa Fluor 680
501137399 565/578 488, 532, 561 PE
501121505 410/455 405 Pacific Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 missing translation for 'options'
Catalog No. A13201 missing translation for 'mfr' Molecular Probes™ missing translation for 'supplierNo' A13201

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.

Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.

Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

Specifications

Description Annexin V, Alexa Fluor 488 conjugate (replacement for Cat. No. PHN1010 and PHN1008)
Quantity 500 μL
Product Type Annexin V conjugate
Content And Storage Contains 1 vial of annexin V, Alexa Fluor 488 conjugate. Store in refrigerator (2°C to 8°C) and protect from light.
Flow Cytometer Laser Lines 488
Excitation/Emission 495/519
No. of Reactions 100
Color Green
Shipping Condition Wet Ice
Conjugate Alexa Fluor 488
For Use With (Equipment) Flow Cytometer
Show More Show Less
I want to study apoptosis using an Annexin V conjugate, but with adherent cells via microscopy instead of flow cytometry. Can this be done?

It has been done, but we don‘t recommend it. Both healthy cells and apoptotic cells possess phosphatidylserine on the cell surface, which can be detected with Annexin V, but apoptotic cells have significantly more of it. You can easily tell the difference between these two populations with flow cytometry, because flow cytometers are more sensitive and have a higher throughput. But with a microscope, you cannot always tell the difference, especially for adherent cells. Instead, for microscopy, we recommend a different technique, such as detecting caspases with CellEvent Caspase Detection Reagents.

I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?

Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Can I detect annexin V staining in an imaging assay?

Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.

When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?

Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Can I fix my cells after annexin V staining?

Yes, this is possible. We have established protocols for annexin V staining combined with intracellular staining of lymphocytes that can be found here. The most important step is to leave some binding buffer in the suspension when fixation is started. Compared to staining of live cells, the intensity of the annexin V signal may be somewhat reduced.

I am trying to label adherent cells with annexin V and am finding that everything is getting labeled. How can I fix this?

Treating cells with trypsin or other reagents to detach adherent cells causes damage to the membrane, such that cells will be labeled with annexin V. The best way to avoid this problem is to allow your cells to recover for 30-45 min in the incubator. Swirl the tube/plate/flask every few minutes to prevent re-attachment. After this recovery period, you can label your cells with annexin V and analyze by flow cytometry.

What are the advantages of flow cytometry?

-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.

What kinds of applications can I run on a flow cytometer?

There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.