missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin Binding Buffer (5X), for flow cytometry

Catalog No. 501121557
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121557 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121557 Supplier Invitrogen™ Supplier No. V13246
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Annexin Binding Buffer (5X), for flow cytometry

This product is designed to facilitate the binding of annexin V to phosphatidylserine for use in apoptosis assays.

Specifications

Quantity 50 mL
pH 7.4
Product Type Annexin Binding Buffer
Content And Storage Contains 1 bottle of annexin binding buffer (5x solution, 50 mL).

Store at 2–8°C.
Shipping Condition Wet Ice
Formulation 50 mM HEPES, 700 mM NaCl, 12.5 mM CaCl2, pH 7.4
For Use With (Application) Flow Cytometry
Form Concentrated
I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?

Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Can I detect annexin V staining in an imaging assay?

Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.

When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?

Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Can I fix my cells after annexin V staining?

Yes, this is possible. We have established protocols for annexin V staining combined with intracellular staining of lymphocytes that can be found here. The most important step is to leave some binding buffer in the suspension when fixation is started. Compared to staining of live cells, the intensity of the annexin V signal may be somewhat reduced.

For Research Use Only. Not for use in diagnostic procedures.