Learn More
Foxx Life Sciences VersaBarb Hose Barb Spigot Fitting, 3/8 Inch, for use with Foxx EZgrip Carboys and Nalgene-Style Containers
Catalog No. 501049859
Encompass
225-4003-RLS: VersaBarb, Spigot Fitting, 1 1/8" Thread with 3/8" HB. Foxx Life Sciences VersaBarb provides flexibility in drainage, pump connection, filling bell, and automated equipment. VersaBarb is available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes. In addition, a 3/4" sanitary fitting VersaBarb is also available to enable the EZgrip carboy to function as a supply reservoir for a larger system. Offers a convenient solution for bottom-connections and can fit a variety of tubing diameters. Made from Class VI autoclavable polypropylene. Available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes or 3/4" sanitary fitting.
Catalog No. 501049859 Supplier Foxx Life Sciences Supplier No. 2254003RLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
225-4003-RLS: VersaBarb, Spigot Fitting, 1 1/8" Thread with 3/8" HB. Foxx Life Sciences VersaBarb provides flexibility in drainage, pump connection, filling bell, and automated equipment. VersaBarb is available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes. In addition, a 3/4" sanitary fitting VersaBarb is also available to enable the EZgrip carboy to function as a supply reservoir for a larger system. Offers a convenient solution for bottom-connections and can fit a variety of tubing diameters. Made from Class VI autoclavable polypropylene. Available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes or 3/4" sanitary fitting.