Learn More
Foxx Life Sciences VersaCap 53mm (53B) Bottle and Flask Caps with Tubing, Open Cap with Tangle Free Hose Barb Adapter, Two 1/8 Inch Barbs with Tubing
Catalog No. 501049842
Encompass
205-6001-RLS: VersaCap, 53mm, Open Cap with 2x 1/8" HB, and Tubing. Foxx Life Sciences Filling and Venting closures are aseptic handling solutions for pharmaceutical, biotechnology, and laboratory liquid transfer applications. All closures can be used on bottles and carboys with 53mm, 83mm, or 120mm closures. The pre-mounted lengths of platinum-cured silicone tubing allow for easy customization of carboys and bottles. Eliminate pouring of high valued products through this leak-proof aseptic system. Flexible system is designed with closures that fit bottles or carboys with 53mm, 83mm, and 120mm closures. Tubing manufactured from high purity silicone. Accept additional fluid transfer components such as aseptic connectors and air vent filters. All components are autoclavable. VersaCap is made from polypropylene, which may be autoclaved at 15 PSI and 121C (250F) for 20 minutes.
Catalog No. 501049842 Supplier Foxx Life Sciences Supplier No. 2056001RLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
205-6001-RLS: VersaCap, 53mm, Open Cap with 2x 1/8" HB, and Tubing. Foxx Life Sciences Filling and Venting closures are aseptic handling solutions for pharmaceutical, biotechnology, and laboratory liquid transfer applications. All closures can be used on bottles and carboys with 53mm, 83mm, or 120mm closures. The pre-mounted lengths of platinum-cured silicone tubing allow for easy customization of carboys and bottles. Eliminate pouring of high valued products through this leak-proof aseptic system. Flexible system is designed with closures that fit bottles or carboys with 53mm, 83mm, and 120mm closures. Tubing manufactured from high purity silicone. Accept additional fluid transfer components such as aseptic connectors and air vent filters. All components are autoclavable. VersaCap is made from polypropylene, which may be autoclaved at 15 PSI and 121C (250F) for 20 minutes.