missing translation for 'onlineSavingsMsg'
Learn More

Anisonitrile, 99%, Thermo Scientific™

Chemical Identifiers

CAS 874-90-8
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001818
InChI Key XDJAAZYHCCRJOK-UHFFFAOYSA-N
Synonym anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
PubChem CID 70129
IUPAC Name 4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158050250
SDS View Documents
Thermo Scientific Chemicals
158050250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC158051000
SDS View Documents
Thermo Scientific Chemicals
158051000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

874-90-8
133.15
XDJAAZYHCCRJOK-UHFFFAOYSA-N
70129
COC1=CC=C(C=C1)C#N
C8H7NO
MFCD00001818
anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
4-methoxybenzonitrile

Specifications

874-90-8
Authentic
Glass bottle
CH3OC6H4CN
25g
anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
XDJAAZYHCCRJOK-UHFFFAOYSA-N
4-methoxybenzonitrile
70129
99%
256°C to 257°C (765.0mmHg)
99%
C8H7NO
MFCD00001818
10, 168
Solubility in water: soluble
COC1=CC=C(C=C1)C#N
133.15
133.15
Anisonitrile, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-871-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only