missing translation for 'onlineSavingsMsg'
Learn More

Anisoin 97.0+%, TCI America™

Catalog No. A101025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A101025G 25 g
A1010100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A101025G Supplier TCI America Supplier No. A101025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-52-8
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD00008411
InChI Key LRRQSCPPOIUNGX-UHFFFAOYNA-N
Synonym anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
PubChem CID 95415
IUPAC Name 2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one
SMILES COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 25 g
Formula Weight 272.30
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Anisoin
EINECSNumber (4)-0277
TSCA Yes