missing translation for 'onlineSavingsMsg'
Learn More

Anisoin, 95%

Catalog No. AC104840250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104840250 25 g Plastic bottle
AC104841000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104840250 Supplier Thermo Scientific Chemicals Supplier No. 104840250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119-52-8 | C16H16O4 | 272.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-52-8
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD00008411
InChI Key LRRQSCPPOIUNGX-UHFFFAOYNA-N
Synonym anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
PubChem CID 95415
IUPAC Name 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 109°C to 114°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 08, 423
Formula Weight 272.29
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Anisoin, 95%
Show More Show Less
EINECSNumber 204-330-8
TSCA TSCA

RUO – Research Use Only