missing translation for 'onlineSavingsMsg'
Learn More

Aniracetam 98.0+%, TCI America™

Catalog No. p-7117878
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23941G 1 g
A23945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23941G Supplier TCI America Supplier No. A23941G
Only null left

Chemical Identifiers

CAS 72432-10-1
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00153767
InChI Key ZXNRTKGTQJPIJK-UHFFFAOYSA-N
Synonym aniracetam, draganon, sarpul, 1-4-methoxybenzoyl-2-pyrrolidinone, ampamet, 1-4-methoxybenzoyl pyrrolidin-2-one, 1-p-anisoyl-2-pyrrolidinone, memodrin, aniracetamun inn-latin, aniracetam usan:inn:jan
PubChem CID 2196
ChEBI CHEBI:47943
IUPAC Name 1-(4-methoxybenzoyl)pyrrolidin-2-one
SMILES COC1=CC=C(C=C1)C(=O)N2CCCC2=O

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Formula Weight 219.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Aniracetam
RTECSNumber UY5781900
TSCA No