Learn More
Oster™ Animal Clipper Size 10 Blade Sets
Supplier: Oster™ OST91904
Includes: As supplied with 01-305-10 animal clippers.
Description
Size 10 Blade Set. For clipping underbody, flanks, head, and face areas. Fine-grain, high-carbon steel. Cuts hair to length of 1/16 in. (1.5mm).
Specifications
| Surgical shaving with 01-305-10 animal clippers | |
| 10 | |
| 1 Ea. |
| Carbon Steel | |
| Clipping underbody, flanks, head and face areas, cuts 1.5 mm |