missing translation for 'onlineSavingsMsg'
Learn More

Aniline, ACS, 99+%

Catalog No. p-3256895
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36238AE 100 mL
AA36238AP 500 mL
AA36238K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36238AE Supplier Thermo Scientific Chemicals Supplier No. 036238AE
Only null left

CAS: 62-53-3 | C6H7N | 93.13 g/mol

It is used to manufacture dyes, photographic chemicals and agricultural chemicals. Its main use is in the manufacture of precursors to polyurethane and other industrial chemicals. Aniline emulsion template is also essential for the synthesis of hollow microspheres of polyaniline. The poly(N-methyl aniline) doped with sulphonic acids plays an important role for humidity sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point °C to 6°C
Density 1.021
Boiling Point 183°C to 184°C
Flash Point 70°C (158°F)
Assay Percent Range 99+%
Refractive Index 1.586
Quantity 100 mL
Beilstein 605631
Solubility Information Slightly soluble in water.
Formula Weight 93.13
Grade ACS
Physical Form Liquid
Chemical Name or Material Aniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H317-H318-H336-H341-H351-H372
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 200-539-3
RTECSNumber BW6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only