missing translation for 'onlineSavingsMsg'
Learn More

Aniline-2,5-disulfonic Acid Monosodium Salt 98.0+%, TCI America™

Catalog No. p-7116795
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A026325G 25 g
A0263500G 500 g
2 options

Catalog No. A026325G

Supplier: TCI America A026325G

Only null left

Chemical Identifiers

CAS 24605-36-5
Molecular Formula C6H5NNa2O6S2
Molecular Weight (g/mol) 297.21
MDL Number MFCD00078393
InChI Key QYUDDCLXBAJUNA-UHFFFAOYSA-L
Synonym 2-Amino-1,4-benzenedisulfonic Acid Monosodium Salt, 2,5-Disulfoaniline Monosodium Salt, Sodium Hydrogen 2-Amino-1,4-benzenedisulfonate
PubChem CID 87094165
IUPAC Name disodium 2-aminobenzene-1,4-disulfonate
SMILES [Na+].[Na+].NC1=CC(=CC=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 275.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Aniline-2,5-disulfonic Acid Monosodium Salt
EINECSNumber (3)-2101&(3)-3169
TSCA No