missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aniline-2,5-disulfonic Acid Monosodium Salt 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 275.22 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Aniline-2,5-disulfonic Acid Monosodium Salt |
Safety and Handling
| EINECSNumber | (3)-2101&(3)-3169 |
| TSCA | No |