missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anhydrous Lactose, NF, Spectrum™ Chemical
LA103, 63-42-3, C12H22O11
$366.42 - $7,632.67
Chemical Identifiers
| CAS | 63-42-3 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.30 |
| InChI Key | GUBGYTABKSRVRQ-UHFFFAOYNA-N |
| IUPAC Name | 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| SMILES | OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600715
|
Spectrum Chemical Mfg Cor
LA103500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18600716
|
Spectrum Chemical Mfg Cor
LA10325KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
|
18600717
|
Spectrum Chemical Mfg Cor
LA10312KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18600718
|
Spectrum Chemical Mfg Cor
LA10345KGBL |
45 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Anhydrous Lactose, NF is used as a tablet diluent in the pharmaceutical industry. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 63-42-3 | |
| 342.30 | |
| 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| C12H22O11 | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O |
Specifications
| 63-42-3 | |
| 0.1% | |
| Poly Bottle | |
| 500 g | |
| GUBGYTABKSRVRQ-UHFFFAOYNA-N | |
| 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
| NF |
| 100% | |
| 0.5% | |
| C12H22O11 | |
| +54.4° to +55.9° | |
| OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O | |
| 342.30 |