missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Angled USB Cable

Catalog No. 14381079
Change view
Click to view available options
:
Angled USB Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14381079 Angled USB Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14381079 Supplier BRANDTECH™ BRAND™ Supplier No. 705240
Only null left

Accessory for use with the HandyStep™ touch

  • USB charging for the HandyStep touch™
  • Angled for use with wall holder part no. 705235

Specifications

Color Black
Disposable Reusable
For Use With BRAND HandyStep Touch/Touch S
Material Plastic
Model USB A/USB C
Number of Channels 1
Type USB Charging Cable
For Use With (Equipment) Electronic Repeating Pipettes
Voltage 4.8 V