missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Angle Rotors

Catalog No. 021061048
Change view
Click to view available options
Capacity:
30 x 1.5/2.0 mL
4 x 8-Place PCR-strips
12 x 5 mL
Net Weight:
0.660 kg
1.560 kg
1.700 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Net Weight
021061048 4 x 8-Place PCR-strips 0.660 kg
021061047 12 x 5 mL 1.560 kg
021061046 30 x 1.5/2.0 mL 1.700 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 021061048 Supplier OHAUS™ Supplier No. 30130874

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

OHAUS™ Angle Rotors for use with Frontier 5000 Series Micro Centrifuge (FC5515)

  • Angle Rotors are available in a variety of different sizes
  • For use with Frontier 5000 Series Centrifuge

Specifications

Description Angle Rotor
Material Stainless Steel
Type Non-sealable
Capacity 4 x 8-Place PCR-strips
Max. RCF 15,342 x g
Max. Speed 15,000 rpm
Net Weight 0.660 kg
For Use With (Equipment) Frontier 5000 Series Micro Centrifuge
Angle 45°
Biocontainment Yes, when used with biocontainment lids