missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Angiotensin II (human)

CAS: 4474-91-31046.197 g/mol

$175.81 - $612.56

Chemical Identifiers

CAS 4474-91-3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60866LB0
View Documents
Thermo Scientific Chemicals
J60866LB0
5 mg
Each for $175.81
Only null left
 
AAJ60866MB
View Documents
Thermo Scientific Chemicals
J60866MB
25 mg
Each for $612.56
Only null left
 
Description

Description

Ang II stimulates angiogenesis and increases microvessel density. In addition, the renin-angiotensin and nitric oxide-generating systems appear to interract in the regulation of cardiovascular function. Angiotensin (Ang) II is a peptide which plays an important role in the regulation of blood pressure and has strong vasoconstrictive effects. It also stimulates the release of aldosterone from the adrenal gland and inhibits adenylate cyclase activity. It also increases the entry of Ca2+ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase (MYLK). It also elicits an increase in voltage-dependent delayed rectifier K+ current in neurons and activates JAK2 in smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ang II stimulates angiogenesis and increases microvessel density. In addition, the renin-angiotensin and nitric oxide-generating systems appear to interract in the regulation of cardiovascular function. Angiotensin (Ang) II is a peptide which plays an important role in the regulation of blood pressure and has strong vasoconstrictive effects. It also stimulates the release of aldosterone from the adrenal gland and inhibits adenylate cyclase activity. It also increases the entry of Ca2+ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase (MYLK). It also elicits an increase in voltage-dependent delayed rectifier K+ current in neurons and activates JAK2 in smooth muscle cells.

Solubility
Soluble in Water (25 mg/ml), and 5% Acetic Acid (1 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. keep away from Strong acids, Strong bases .
Specifications

Chemical Identifiers

4474-91-3

Specifications

4474-91-3
Store at -20°C
1046.197
5 mg
White
Powder
Angiotensin II (human)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only