Recombinant Protein;200UG;Biotinylated Human Angiopoietin-2, His,Avitag (AN2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # AAI26201.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 29.2 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.