missing translation for 'onlineSavingsMsg'
Learn More

Angelicin 98.0+%, TCI America™

Supplier:  TCI America A257325MG

Catalog No. A257325MG

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Angelicin
140°C
104°C
MFCD00064930
Isopsoralen, 2H-Furo[2,3-h]chromen-2-one, Furo[2,3-h]benzopyran-2-one
C1=CC2=C(C=CO2)C3=C1C=CC(=O)O3
186.166
CHEBI:28928
≥98.0% (GC)
523-50-2
White-Yellow
C11H6O3
25 mg
XDROKJSWHURZGO-UHFFFAOYSA-N
furo[2,3-h]chromen-2-one
10658
186.17
Crystalline Powder

Chemical Identifiers

523-50-2
186.166
XDROKJSWHURZGO-UHFFFAOYSA-N
10658
furo[2,3-h]chromen-2-one
C11H6O3
MFCD00064930
Isopsoralen, 2H-Furo[2,3-h]chromen-2-one, Furo[2,3-h]benzopyran-2-one
CHEBI:28928
C1=CC2=C(C=CO2)C3=C1C=CC(=O)O3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : LV0940000

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
Documents

Documents

Promotions

Promotions