missing translation for 'onlineSavingsMsg'
Learn More

Angelic Acid 98.0+%, TCI America™

Catalog No. p-7117262
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11361G 1 g
A11365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11361G Supplier TCI America Supplier No. A11361G

Chemical Identifiers

CAS 565-63-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00002654
InChI Key UIERETOOQGIECD-ARJAWSKDSA-N
Synonym (Z)-2-Methyl-2-butenoic Acid, 2-Methylisocrotonic Acid
PubChem CID 643915
ChEBI CHEBI:36431
IUPAC Name (Z)-2-methylbut-2-enoic acid
SMILES CC=C(C)C(=O)O

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 96°C
Quantity 1 g
UN Number 3261
Formula Weight 100.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Angelic Acid
EINECSNumber (2)-0609
TSCA No
Recommended Storage Refrigerator