missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corepoint Scientific™ Anchor Brackets
Description
- Anchor bracket kit for anti-tip security
- Designed for Corepoint Scientific™ Blood Bank Refrigerators and Plasma Freezers
Specifications
Specifications
| Cabinet Material | Stainless Steel |
| Color | Silver |
| For Use With (Application) | Anchor bracket kit to provide anti-tip resistance |
| For Use With (Equipment) | Corepoint™ Blood Bank Refrigerators and Plasma Freezers |
| Includes | 2 Brackets |
| Model | ANCHOR KIT |