Learn More
Organomation ANALYTICAL NITROGEN EVAPORATOR DFS Item
Supplier:  Organomation 11250RT
This evaporator is acid resistant and capable of processing 24 samples at a time and provides controlled concentration of sample vials through the application of nitrogen, within a water bath, with individualized control of each sample
Catalog No. NC9892499
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions