Learn More
Scientific Industries Inc BALANCE, ANALYTICAL, AUTO CAL DFS Item
Supplier:  Scientific Industries Inc AGCN120
Analytical Balance, 120g x 0.0001g w/ Automatic Internal Calibration keeping balance calibrated at all times, includes 5 weighing functions, a large glass weighing chamber, die-cast metal housing, and equipped with RS232, USB, Clock, Backlit LCD Display.
Catalog No. 501139354
Only null left
Explore available promotions