Learn More
Thermo Scientific™ AnaeroPack™-Anaero Anaerobic Gas Generator
Catalog No. 23246376
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
23246376 20/Pk.
1 options
Catalog No. 23246376 Supplier Thermo Scientific™ Supplier No. NRE4650
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mitsubishi™ AnaeroPack-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
Mitsubishi™ AnaeroPack-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
  • No hydrogen gas produced
  • Use 1 sachet/2.5L jar or 3 sachets/7.0L jar
Specifications
Type AnaeroPack-Anaero
Format Gas Generation Sachet
Product Line Thermo Scientific Mitsubishi
Quantity 20/Pk.