missing translation for 'onlineSavingsMsg'
Learn More

Interscience Anabac

Catalog No. 16948946
Change view
Click to view available options
Scent:
Apple
Coconut
Eucalyptus
Floral
Lemon
Orchard Peach
Vanilla
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent
16948946 Apple
16948941 Eucalyptus
16948940 Lemon
16948945 Orchard Peach
16948942 Coconut
16948944 Vanilla
16948943 Floral
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 16948946 Supplier Interscience Supplier No. 320200
Only null left

Autoclave deodorant. Available in a variety of scents.

Autoclave deodorant available in the following scents:

  • Eucalyptus
  • Apple
  • Lemon
  • Orchard Peach
  • Coconut, 100% natural perfume, Ecocert Certified
  • Vanilla, 100% natural perfume, Ecocert Certified
  • Floral, 100% natural perfume, Ecocert Certified

Specifications

Product Type Autoclave Capsule
Color-coded Yes
Form Capsules
Scent Apple
Sterility Non-sterile
Appearance Clear
Autoclavable Autoclavable
Color Green
Disposable Single-use
Filtered Non-filtered
Flammable No
For Use With (Application) Autoclave Deodorant
Quantity 100/Cs.
Show More Show Less