Learn More
New England Biolab Ltd Amylose Resin 15 ml
Supplier:  New England Biolab Ltd E8021S
Encompass
Amylose resin is an affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP). It is intended for use in a gravity flow column.
Catalog No. 50811582
Only null left