missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amylopectin, MP Biomedicals™
$409.61 - $409.61
Chemical Identifiers
| CAS | 9037-22-3 |
|---|---|
| Molecular Formula | C30H52O26 |
| Molecular Weight (g/mol) | 828.72 |
| MDL Number | MFCD00130510 |
| InChI Key | WMGFVAGNIYUEEP-WUYNJSITSA-N |
| Synonym | amylopectin, amylopectine, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl tetrahydropyran-3-yl oxy-tetrahydropyran-2-yl meth, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-2-2s,3r,4r,5s,6r-3,4-dihydroxy-6-hydroxymethyl-5-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxan-2-yl oxy methyl-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
| PubChem CID | 439207 |
| ChEBI | CHEBI:28057 |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-4,5-dihydroxy-2-(hydroxymethy |
| SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OCC3C(C(C(C(O3)OC4C(OC(C(C4O)O)O)CO)O)O)OC5C(C(C(C(O5)CO)O)O)O)CO)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19504891
|
MP Biomedicals Inc
219504891 |
1 kg |
Each for $409.61
|
|
|||||
Chemical Identifiers
| 9037-22-3 | |
| 828.72 | |
| WMGFVAGNIYUEEP-WUYNJSITSA-N | |
| 439207 | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-4,5-dihydroxy-2-(hydroxymethy |
| C30H52O26 | |
| MFCD00130510 | |
| amylopectin, amylopectine, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl tetrahydropyran-3-yl oxy-tetrahydropyran-2-yl meth, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-2-2s,3r,4r,5s,6r-3,4-dihydroxy-6-hydroxymethyl-5-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxan-2-yl oxy methyl-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| CHEBI:28057 | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OCC3C(C(C(C(O3)OC4C(OC(C(C4O)O)O)CO)O)O)OC5C(C(C(C(O5)CO)O)O)O)CO)O)O)O)O |
Specifications
| White | |
| 9037-22-3 | |
| MFCD00130510 | |
| 1 kg | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OCC3C(C(C(C(O3)OC4C(OC(C(C4O)O)O)CO)O)O)OC5C(C(C(C(O5)CO)O)O)O)CO)O)O)O)O | |
| 828.72 | |
| CHEBI:28057 |
| Powder | |
| C30H52O26 | |
| amylopectin, amylopectine, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl tetrahydropyran-3-yl oxy-tetrahydropyran-2-yl meth, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-2-2s,3r,4r,5s,6r-3,4-dihydroxy-6-hydroxymethyl-5-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxan-2-yl oxy methyl-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| WMGFVAGNIYUEEP-WUYNJSITSA-N | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-4,5-dihydroxy-2-(hydroxymethy | |
| 439207 | |
| Amylopectin, from Corn |
Safety and Handling
EINECSNumber : 232-911-6
Recommended Storage : Store at Room Temperature (15-30°C).