missing translation for 'onlineSavingsMsg'
Learn More

Amylopectin Hydrate (Amylose free), from Waxy Corn, TCI America™

Catalog No. A0456500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0456500G 500 g
A045625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0456500G Supplier TCI America Supplier No. A0456500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9037-22-3
Molecular Formula C30H52O26
Molecular Weight (g/mol) 828.72
MDL Number MFCD00130510
InChI Key WMGFVAGNIYUEEP-WUYNJSITSA-N
Synonym amylopectin, amylopectine, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl tetrahydropyran-3-yl oxy-tetrahydropyran-2-yl meth, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-2-2s,3r,4r,5s,6r-3,4-dihydroxy-6-hydroxymethyl-5-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxan-2-yl oxy methyl-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439207
ChEBI CHEBI:28057
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-4,5-dihydroxy-2-(hydroxymethy
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OCC3C(C(C(C(O3)OC4C(OC(C(C4O)O)O)CO)O)O)OC5C(C(C(C(O5)CO)O)O)O)CO)O)O)O)O

Specifications

Color White
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Amylopectin Hydrate (Amylose free), from Waxy Corn