missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amyloid beta (1-42), Human

Catalog No. AAJ66387MCR
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66387MCR 1 mg
AAJ66387EXD 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66387MCR Supplier Thermo Scientific Chemicals Supplier No. J66387MCR

For cellular and molecular biology applications | 4514.03 g/mol

Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer’s disease and neurodegeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
For Use With (Application) Insoluble in water. See J66591 for water soluble hydrochloride form
Molecular Weight (g/mol) 4514.03
Product Type Amyloid beta (1-42), human
Quantity 1 mg
Regulatory Status RUO - research use only

RUO – Research Use Only