missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amylin, Human

Catalog No. AAJ66984MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66984MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66984MCR Supplier Thermo Scientific Chemicals Supplier No. J66984MCR

 

Amylin functions as part of the endocrine pancreas and contributes to glycemic control. It functions as a synergistic partner to insulin. The overall effect is to slow the rate of appearance (Ra) of glucose in the blood after eating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 3903.27
Product Type Amylin, human
Quantity 1 mg
Regulatory Status RUO

RUO – Research Use Only