missing translation for 'onlineSavingsMsg'
Learn More

Amylcyclohexane 98.0+%, TCI America™

Catalog No. A074525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A074525ML Supplier TCI America Supplier No. A074525ML

Chemical Identifiers

CAS 4292-92-6
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00019402
InChI Key HLTMUYBTNSVOFY-UHFFFAOYSA-N
Synonym n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl
PubChem CID 20284
IUPAC Name pentylcyclohexane
SMILES CCCCCC1CCCCC1

Specifications

Melting Point -58°C
Color Colorless
Boiling Point 202°C
Quantity 25 mL
UN Number 3082
Formula Weight 154.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amylcyclohexane