missing translation for 'onlineSavingsMsg'
Learn More

Amylbenzene 98.0+%, TCI America™

Catalog No. A044925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A044925ML 25 mL
A0449500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A044925ML Supplier TCI America Supplier No. A044925ML

Chemical Identifiers

CAS 538-68-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00009502
InChI Key PWATWSYOIIXYMA-UHFFFAOYSA-N
Synonym 1-phenylpentane, n-pentylbenzene, amylbenzene, n-amylbenzene, benzene, pentyl, 1-pentylbenzene, 1-phenyl-n-pentane, pentane, 1-phenyl, pentyl-benzene, pentane, phenyl
PubChem CID 10864
IUPAC Name pentylbenzene
SMILES CCCCCC1=CC=CC=C1

Specifications

Melting Point -75°C
Color Colorless
Boiling Point 205°C
Quantity 25 mL
UN Number 3082
Formula Weight 148.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amylbenzene
EINECSNumber (3)-0021
TSCA Yes