missing translation for 'onlineSavingsMsg'
Learn More

Amyl Nitrate 98.0+%, TCI America™

Catalog No. A045325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A045325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A045325ML Supplier TCI America Supplier No. A045325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1002-16-0
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00059177
InChI Key HSNWZBCBUUSSQD-UHFFFAOYSA-N
Synonym Pentyl Nitrate
PubChem CID 61250
IUPAC Name pentyl nitrate
SMILES CCCCCO[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 157°C
Quantity 25 mL
UN Number 1112
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Nitrate
EINECSNumber (2)-1581
RTECSNumber QV0600000
TSCA No