missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl Butyrate 98.0+%, TCI America™
$35.41 - $236.73
Chemical Identifiers
| CAS | 540-18-1 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.24 |
| MDL Number | MFCD00048850 |
| InChI Key | CFNJLPHOBMVMNS-UHFFFAOYSA-N |
| Synonym | pentyl butyrate, amyl butyrate, n-amyl butyrate, butanoic acid, pentyl ester, amyl butanoate, 1-pentyl butyrate, n-pentyl butanoate, n-pentyl n-butyrate, n-pentyl butyrate, butyric acid, pentyl ester |
| PubChem CID | 10890 |
| ChEBI | CHEBI:87684 |
| IUPAC Name | pentyl butanoate |
| SMILES | CCCCCOC(=O)CCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B075525ML
|
TCI America
B075525ML |
25 mL |
Each for $35.41
|
|
|||||
|
B0755500ML
|
TCI America
B0755500ML |
500 mL |
Each for $236.73
|
|
|||||
Chemical Identifiers
| 540-18-1 | |
| 158.24 | |
| CFNJLPHOBMVMNS-UHFFFAOYSA-N | |
| 10890 | |
| pentyl butanoate |
| C9H18O2 | |
| MFCD00048850 | |
| pentyl butyrate, amyl butyrate, n-amyl butyrate, butanoic acid, pentyl ester, amyl butanoate, 1-pentyl butyrate, n-pentyl butanoate, n-pentyl n-butyrate, n-pentyl butyrate, butyric acid, pentyl ester | |
| CHEBI:87684 | |
| CCCCCOC(=O)CCC |
Specifications
| 540-18-1 | |
| 185°C | |
| MFCD00048850 | |
| 2620 | |
| CFNJLPHOBMVMNS-UHFFFAOYSA-N | |
| pentyl butanoate | |
| 10890 | |
| 158.24 | |
| Liquid |
| Colorless | |
| C9H18O2 | |
| 25 mL | |
| pentyl butyrate, amyl butyrate, n-amyl butyrate, butanoic acid, pentyl ester, amyl butanoate, 1-pentyl butyrate, n-pentyl butanoate, n-pentyl n-butyrate, n-pentyl butyrate, butyric acid, pentyl ester | |
| CCCCCOC(=O)CCC | |
| 158.24 | |
| CHEBI:87684 | |
| ≥98.0% (GC) | |
| Amyl Butyrate |
Safety and Handling
EINECSNumber : (2)-0779
RTECSNumber : ET5956000
TSCA : Yes