missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetoacetate 98.0+%, TCI America™

Catalog No. A0811500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0811500ML 500 mL
A081125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0811500ML Supplier TCI America Supplier No. A0811500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6624-84-6
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00059433
InChI Key IDZAUPYMMSSVHP-UHFFFAOYSA-N
Synonym Acetoacetic Acid Pentyl Ester, Acetoacetic Acid Amyl Ester, Pentyl Acetoacetate
PubChem CID 138786
IUPAC Name pentyl 3-oxobutanoate
SMILES CCCCCOC(=O)CC(C)=O

Specifications

Color Yellow
Boiling Point 222°C
Quantity 500 mL
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Acetoacetate
EINECSNumber (2)-1505
TSCA No