missing translation for 'onlineSavingsMsg'
Learn More

Amygdalin 97.0+%, TCI America™

Catalog No. A04435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04435G 5 g
A04431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04435G Supplier TCI America Supplier No. A04435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29883-15-6
Molecular Formula C20H27NO11
Molecular Weight (g/mol) 457.43
MDL Number MFCD29049822
InChI Key XUCIJNAGGSZNQT-ARGBPYSKNA-N
Synonym amygdalin
PubChem CID 66509032
IUPAC Name 2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile
SMILES OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 213°C
Color White
Quantity 5 g
Formula Weight 457.43
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Amygdalin
RTECSNumber OO8450000
TSCA No