Learn More
Agilent BioTek AMX Cassette
GREENER_CHOICE
Catalog No. BT1260035
Change view
Click to view available options
For Use With (Equipment):
MFPX2
Catalog No. For Use With (Equipment)
BT1260035 MFPX2
1 options
Catalog No. BT1260035 Supplier Agilent BioTek Supplier No. 1260035
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Two cassettes are required for use with the AMX module (PN 1260041). User installs cassettes with AMX manifolds (included with AMX module). For use with MFXP2R, order two cassettes (does not require PN 1260041).
Dispensing speeds:
  • 96 wells, 5 μL cass, 10 μL/ well: 3 seconds
  • 96 wells, 5 μL cass, 20 μL/ well: 3.5 seconds
  • 384 wells, 5 μL cass, 5 μL/ well: 6.5 seconds
  • 384 wells, 10 μL cass, 10 μL/ well: 8 seconds
  • 384 wells, 1 μL cass, 1 μL/ well: 6 seconds
  • 1536 wells, 1 μL cass, 1 μL/ well: 21 seconds
Specifications
Accuracy <5%
Autoclavable Autoclavable
For Use With (Application) 2 D and 3 D Cell Culture to Concentration Normalization Assays, ELISA, Bead-Based Assays and More
For Use With (Equipment) MFPX2
Includes Two 8-channel autoclavable manifolds, tube-in-tube design
Precision <5% CV
Electrical Requirements 50/60 Hz