Learn More
Beckman Coulter AMPure XP, 5 mL
Supplier: Beckman Coulter A63880
A highly efficient, easily automated PCR purification system that delivers superior quality DNA with no salt carryover. Requiring no centrifugation or filtration, AMPure XP can be easily used in manual and automated 96- or 384-well formats