missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ampule Breakers

Thermo Scientific Ampule Breakers provide a safe, convenient way to open 1 mL to 10 mL glass ampules.

Supplier:  Thermo Scientific™ 28915

Encompass

 View more versions of this product

Catalog No. 28915


Only null left

Description

Description

These single-use polyethylene sleeve ampule breakers help protect your fingers from glass shards when opening 1 mL to 10 mL glass ampules. The Thermo Scientific Ampule Breakers encapsulate the ampule top upon opening, making disposal of the glass top easy and safe.

  • Easy to use single-use format
  • Compatible with 1 mL to 10 mL glass ampules
  • Protects against glass shards
  • Helps prevent cross contamination
Specifications

Chemical Identifiers

Single-use
Ampule Breaker

Specifications

Single-use
100 Breakers
100 breakers
Ampule Breaker
1-4 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions