Learn More
LGC Genomics Llc (Lucigen) AmpliScribe T7-Flash Transcription Kit, 25 RxnsProduce the highest yields of RNA transcript in a fast, scalable 30-minute protocol.
Catalog No. NC1736578
Encompass
The AmpliScribe™ T7-Flash™ Transcription Kit produces more RNA in 30 minutes than other in vitro transcription kits produce in two hours.
Catalog No. NC1736578 Supplier LGC Genomics Llc Supplier No. ASF3257
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The AmpliScribe™ T7-Flash™ Transcription Kit produces more RNA in 30 minutes than other in vitro transcription kits produce in two hours.