Learn More
Gilson™ AmpliPurTM Expert Tips
AmpliPurTM Expert Tips are ISO 8655 compliant and provide a significant value for the money spent. These filter tips protect precious samples and your pipettes from cross-contamination and are designed to fit various pipette brands.
Supplier:  Gilson™ F174201
 View more versions of this product
Catalog No. F174201G
Only null left
Description
Description
  • AmpliPur Expert Tips are designed to fit various pipette brands for maximum convenience on the bench.
  • Equipped with a filter, they also help prevent cross-contamination.
  • Ideal for Molecular Biology.
  • These tips significantly contribute to molecular testing performance by ensuring sample integrity during the pipetting process.
  • Manufactured according to the certified Gilson quality standard, AmpliPur Expert Tips comply with ISO 8655 requirements and are certified free of RNase and DNase, preserving your sample integrity.
Specifications
Specifications
10 to 20 μL
Non-sterile
Yellow
2 in.
ISO 8655
Molecular Biology
Rectangular Prism
960/Pk.
Yellow
Universal Fit
AmpliPur
Filtered
Clear
Not Autoclavable
52 mm
Yes
Rack
Tipack
Air Displacement Tips
Single-use
No
Polypropylene
Product Suggestions
Product Suggestions