Learn More
Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate
Available with black alpha-numeric lettering. Amplate™ T323 Series Thin Wall PCR Plate, each well features ultrathin sidewalls for uniform and efficient temperature transfer.
$93.39 - $93.39
Specifications
| Format | Standard |
|---|---|
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Sterility | Non-sterile |
| No. per Case | 50/Cs. |
| Catalog Number | Mfr. No. | Color | No. of Wells | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | No. of Wells | Price | Quantity | ||||
|
23-045-243
|
Simport™ Scientific
T32324N |
Natural | 24 |
Case of 50 Each for $93.39
|
|
||||
Description
- Made of polypropylene
- Available with black alpha-numeric
- Flexible, can be cut to desired format
- Autoclavable
- Inside of tube is smooth and has an inert surface
Certifications
RNase, DNase, Pyrogen and DNA-free.
Specifications
| Standard | |
| Polypropylene | |
| 50/Cs. |
| Autoclavable | |
| Non-sterile |