Learn More
Thermo Scientific Chemicals Ampicillin Sodium Salt
CAS: 69-52-3 | C16H18N3NaO4S | 371.39 g/mol
$170.85 - $592.33
Chemical Identifiers
| CAS | 69-52-3 |
|---|---|
| Molecular Formula | C16H18N3NaO4S |
| Molecular Weight (g/mol) | 371.39 |
| MDL Number | MFCD00064313 |
| InChI Key | BSFVNXCYXDYHOD-ZQDFAFASSA-N |
| PubChem CID | 131673879 |
| IUPAC Name | (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
| SMILES | [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC611770050
|
Thermo Scientific Chemicals
611770050 |
5 g |
Each for $170.85
|
|
|||||
|
AC611770250
|
Thermo Scientific Chemicals
611770250 |
25 g |
Each for $592.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 69-52-3 | |
| 371.39 | |
| BSFVNXCYXDYHOD-ZQDFAFASSA-N | |
| (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
| C16H18N3NaO4S | |
| MFCD00064313 | |
| 131673879 | |
| [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
Specifications
| Ampicillin Sodium Salt | |
| 8.0 to 10 | |
| C16H18N3NaO4S | |
| 15, 583 | |
| [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] | |
| 371.39 | |
| 371.39 |
| 69-52-3 | |
| 845 - 988 MCG/MG | |
| MFCD00064313 | |
| BSFVNXCYXDYHOD-ZQDFAFASSA-N | |
| (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium | |
| 131673879 | |
| Ampicillin sodium salt |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 200-708-1
RTECSNumber : XH8400000
RUO – Research Use Only