missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphotericin B, Antibiotic Solution, MP Biomedicals
Antibiotic solution Concentration: 250μg/mL Effective against yeast and fungi
$85.57 - $212.33
Chemical Identifiers
| CAS | 1397-89-3 |
|---|---|
| Molecular Formula | C47H73NO17 |
| Molecular Weight (g/mol) | 924.09 |
| MDL Number | MFCD00877763 |
| InChI Key | APKFDSVGJQXUKY-ZNVUZQDLSA-N |
| Synonym | Fungizone |
| PubChem CID | 134129663 |
| IUPAC Name | (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid |
| SMILES | C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Price | Quantity & Availability | |||||
|
ICN1672346
|
N/A | 20 mL |
Each for $85.57
|
|
|||||
|
ICN1672348
|
N/A | 50 mL |
Each for $212.33
|
|
|||||
Chemical Identifiers
| 1397-89-3 | |
| 924.09 | |
| APKFDSVGJQXUKY-ZNVUZQDLSA-N | |
| 134129663 | |
| C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
| C47H73NO17 | |
| MFCD00877763 | |
| Fungizone | |
| (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid |
Specifications
| Amphotericin B | |
| C47H73NO17 | |
| Fungizone | |
| C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O | |
| 924.09 | |
| Amphotericin B |
| 1397-89-3 | |
| MFCD00877763 | |
| APKFDSVGJQXUKY-ZNVUZQDLSA-N | |
| (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid | |
| 134129663 |