missing translation for 'onlineSavingsMsg'
Learn More

Thermo Fisher Scientific Amphipol A8-35

Catalog No. FERA50943
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA50943 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA50943 Supplier Thermo Fisher Scientific Supplier No. A50943

An amphipathic polymer ideally suited to improve solubilization and stabilization of membrane proteins while maintaining structural integrity and activity.

  • An amphipathic polymer ideally suited to improve solubilization and stabilization of membrane proteins while maintaining structural integrity and activity
  • Detergent-free substitute to extract and stabilize membrane proteins in aqueous solutions
  • Surfactant with dense distribution of hydrophobic chains that have a high affinity for transmembrane surfaces
  • Enables protein solubilization with minimal disruption of membrane environment
  • Facilitates membrane protein structure/folding
  • Suitable for protein solubilization and stabilization for single particle cryo-EM analysis

Specifications

Color White
Molecular Formula (C6.2H10.3O1.35N0.65Na0.35)∼72
Quantity 100 mg
Solubility Information Up to 20% in water
Formula Weight 9 kDa
Physical Form Solid