missing translation for 'onlineSavingsMsg'
Learn More

Ampac™ Flexibles StandPAK Pouches

Heat sealable 4.5mL stand-up pouch with tear notches

Supplier:  Ampac™ KAPSP6801EZ500

Catalog No. 01-812-74


Only null left
Explore available promotions

Description

Description

  • Perfect for medical, environmental, and industrial purposes
  • Withstand boiling and freezing
  • Can be used for long-term specimen/evidence storage

Specifications

Specifications

4.5 mL
9 x 6 in. (22.9 x 58.1cm)
22.9 cm
58.1 cm
4 cm
Polyester
9 in.
6 in.
1.563 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions