Learn More
Ampac™ Flexibles Silver Pak Bags
The 600 Series metalized pouches are ideal for powders and other granular materials
Supplier: Ampac™ 602B1M
Description
SilverPAK ofers a 2-ply laminated structure consisting of metalized polyester and polyethylene (METPET/PE).
Chemical Identifiers
| 2.50 mil |
Specifications
| 2.50 mil | |
| 8 in. | |
| 6.5 in. |