missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin trihydrate

Catalog No. p-7042081
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6129006 5 g
AAJ6129014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6129006 Supplier Thermo Scientific Chemicals Supplier No. J6129006
Only null left

A semi-synthetic antibiotic similar to penicillin | CAS: 61336-70-7 | C16H25N3O8S | 419.449 g/mol

Amoxicillin trihydratesemi-synthetic antibiotic related to Penicillin which has antibacterial poperties. It is used for treatment of a variety of bacterial infections. It is used as a pharmaceutical product as a antibiotic agent, anti-infective drug raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amoxicillin trihydratesemi-synthetic antibiotic related to Penicillin which has antibacterial poperties. It is used for treatment of a variety of bacterial infections. It is used as a pharmaceutical product as a antibiotic agent, anti-infective drug raw materials.

Solubility
Soluble in water at 3.4mg/ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C

Chemical Identifiers

CAS 61336-70-7
Molecular Formula C16H25N3O8S
Molecular Weight (g/mol) 419.449
MDL Number MFCD00072029
InChI Key MQXQVCLAUDMCEF-FXHJIJKUSA-N
PubChem CID 124080955
IUPAC Name (2S,5R,6S)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C.O.O.O

Specifications

Type Amoxicillin Trihydrate
Physical Form Powder
Color White
Quantity 5 g
Beilstein 7507120
Merck Index 14,577
Solubility Information Soluble in water at 3;4mg/ml.
Formula Weight 419.45 (365.40 Anhydrous)
Chemical Name or Material Amoxicillin trihydrate
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 243-003-8
RTECSNumber XH8310000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only